0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Proteins and Peptides
Recombinant Mouse CXCL5 (from HEK293 cells)
Recombinant Mouse CXCL5 (from HEK293 cells)
ABNOP7466.10UG
:
Abnova
:
Recombinant Mouse CXCL5 (from HEK293 cells)
ABNOP7466.10UG
:
Abnova
:
Pk:
10 µG
Protein/peptide type:
Recombinant
Source:
HEK293 cells
Species:
Mouse
Gene ID:
P50228
Protein/peptide name:
CXCL5
Sequence:
45a.a.-118a.a.