Recombinant Human SHH (from E. coli)
Catalog # ABNOP7255.25UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:25 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:Q15465
- Protein/peptide name:SHH
- Sequence:22a.a.-197a.a.