Order Entry
Austria
ContactUsLinkComponent
Recombinant Human PPBP (from E. coli)
Recombinant Human PPBP (from E. coli)
Catalog # ABNOP7201.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Human PPBP (from E. coli)
Catalog # ABNOP7201.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    P02775
  • Protein/peptide name:
    PPBP
  • Sequence:
    35a.a.-107a.a.

Specifications

About this item