Order Entry
Austria
ContactUsLinkComponent
Tamper Evident Closure for Glass Bottles
Tamper Evident Closure for Glass Bottles
Catalog # RIXI8-1100-45-BL
Supplier:  RIXIUS
Tamper Evident Closure for Glass Bottles
Catalog # RIXI8-1100-45-BL
Supplier:  RIXIUS

Specifications

  • Thread:
    GL 45
  • Colour:
    Blue
  • Description:
    Tamper-evident closure for glass bottles, 16,87 g
  • Length:
    35 mm
  • Material:
    PP
  • Pk:
    25

Specifications

About this item