Anti-MCAM Rabbit Recombinant Antibody [clone: 8A10]
ABNORAB04174.100UL
: Abnova
- Antigen symbol:MCAM
- Clonality:Recombinant
- Clone:8A10
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human MCAM.
- Pk:100 µl