Anti-COX1 Rabbit Recombinant Antibody [clone: R02-6G4]
ABNORAB01494.100UL
: Abnova
- Antigen symbol:COX1
- Clonality:Recombinant
- Clone:R02-6G4
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human COX1.
- Pk:100 µl