Order Entry
Austria
ContactUsLinkComponent
 
Anti-Claudin 3 Rabbit Recombinant Antibody [clone: R08-4F8]
  ABNORAB01500.100UL
 :  Abnova
Anti-Claudin 3 Rabbit Recombinant Antibody [clone: R08-4F8]
  ABNORAB01500.100UL
 :  Abnova

 

  • Antigen symbol:
    Claudin 3
  • Clonality:
    Recombinant
  • Clone:
    R08-4F8
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Claudin 3.
  • Pk:
    100 µl