Order Entry
Austria
ContactUsLinkComponent
Anti-CDKN2A/p16INK4a Rabbit Recombinant Antibody [clone: R02-6H1]
Anti-CDKN2A/p16INK4a Rabbit Recombinant Antibody [clone: R02-6H1]
Catalog # ABNORAB01508.100UL
Supplier:  Abnova
Anti-CDKN2A/p16INK4a Rabbit Recombinant Antibody [clone: R02-6H1]
Catalog # ABNORAB01508.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    CDKN2A/p16INK4a
  • Clonality:
    Recombinant
  • Clone:
    R02-6H1
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human CDKN2A/p16INK4a.
  • Pk:
    100 µl

Specifications

About this item