Order Entry
Austria
ContactUsLinkComponent
Anti-CDKN2A/p16INK4a Rabbit Recombinant Antibody [clone: R04-8C9]
Anti-CDKN2A/p16INK4a Rabbit Recombinant Antibody [clone: R04-8C9]
  ABNORAB01510.100UL
 :  Abnova
Anti-CDKN2A/p16INK4a Rabbit Recombinant Antibody [clone: R04-8C9]
  ABNORAB01510.100UL
 :  Abnova

 

  • Antigen symbol:
    CDKN2A/p16INK4a
  • Clonality:
    Recombinant
  • Clone:
    R04-8C9
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human CDKN2A/p16INK4a.
  • Pk:
    100 µl