Anti-CDK6 Rabbit Recombinant Antibody [clone: R03-2G5]
ABNORAB01512.100UL
: Abnova
- Antigen symbol:CDK6
- Clonality:Recombinant
- Clone:R03-2G5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Cdk6.
- Pk:100 µl