Anti-CD55 Rabbit Recombinant Antibody [clone: R01-7H4]
Catalog # ABNORAB01521.100UL
Supplier: Abnova
Specifications
- Antigen symbol:CD55
- Clonality:Recombinant
- Clone:R01-7H4
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human CD55.
- Pk:100 µl