Anti-Bad Rabbit Recombinant Antibody [clone: R07-1B6]
Catalog # ABNORAB01571.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Bad
- Clonality:Recombinant
- Clone:R07-1B6
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Bad.
- Pk:100 µl