Anti-Caspase-3 p12 Rabbit Recombinant Antibody [clone: R05-2D6]
Catalog # ABNORAB01538.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Caspase-3 p12
- Clonality:Recombinant
- Clone:R05-2D6
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human Caspase-3 p12
- Pk:100 µl