Anti-Caspase-1 Rabbit Recombinant Antibody [clone: R05-7H1]
Catalog # ABNORAB01539.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Caspase-1
- Clonality:Recombinant
- Clone:R05-7H1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human Caspase-1
- Pk:100 µl