Anti-Calnexin Rabbit Recombinant Antibody [clone: R07-2B1]
ABNORAB01545.100UL
: Abnova
- Antigen symbol:Calnexin
- Clonality:Recombinant
- Clone:R07-2B1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Calnexin.
- Pk:100 µl