Anti-Caldesmon Rabbit Recombinant Antibody [clone: R05-3D2]
ABNORAB01546.100UL
: Abnova
- Antigen symbol:Caldesmon
- Clonality:Recombinant
- Clone:R05-3D2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Caldesmon.
- Pk:100 µl