Anti-BTF3 Rabbit Recombinant Antibody [clone: R01-2B7]
Catalog # ABNORAB01549.100UL
Supplier: Abnova
Specifications
- Antigen symbol:BTF3
- Clonality:Recombinant
- Clone:R01-2B7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human BTF3
- Pk:100 µl