Anti-BRAT1 Rabbit Recombinant Antibody [clone: R02-4D9]
ABNORAB01551.100UL
: Abnova
- Antigen symbol:BRAT1
- Clonality:Recombinant
- Clone:R02-4D9
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human BRAT1
- Pk:100 µl