Order Entry
Austria
ContactUsLinkComponent
 
Anti-beta 2 Microglobulin Rabbit Recombinant Antibody [clone: R07-8B6]
  ABNORAB01563.100UL
 :  Abnova
Anti-beta 2 Microglobulin Rabbit Recombinant Antibody [clone: R07-8B6]
  ABNORAB01563.100UL
 :  Abnova

 

  • Antigen symbol:
    beta 2 Microglobulin
  • Clonality:
    Recombinant
  • Clone:
    R07-8B6
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human beta 2 Microglobulin.
  • Pk:
    100 µl