Anti-EIF4B Rabbit Recombinant Antibody [clone: R05-4B2]
ABNORAB01788.100UL
: Abnova
- Antigen symbol:EIF4B
- Clonality:Recombinant
- Clone:R05-4B2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human EIF4B.
- Pk:100 µl