Anti-SURF4 Rabbit Recombinant Antibody [clone: R07-1F1]
Catalog # ABNORAB01813.100UL
Supplier: Abnova
Specifications
- Antigen symbol:SURF4
- Clonality:Recombinant
- Clone:R07-1F1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human SURF4.
- Pk:100 µl