Anti-CLDN6 Mouse Recombinant Antibody [clone: 4H4]
Catalog # ABNOH00009074-M02
Supplier: Abnova
Specifications
- Antigen symbol:CLDN6
- Clonality:Recombinant
- Clone:4H4
- Host:Mouse
- Immunogen:Mouse monoclonal antibody raised against a full-length recombinant CLDN6.
- Pk:100 µG