Anti-CHEK1 (pSer345) Rabbit Monoclonal Antibody [clone: R3F9] (FITC)
Artikelnr. ABNOMAB23461.100
Lieferant: Abnova
Spezifikationen
- Antigen symbol:CHEK1
- Clonality:Monoclonal
- Clone:R3F9
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Reactivity:Human
- Modification:pSer345
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human CHEK1
- Pk:100 AU