Order Entry
Austria
ContactUsLinkComponent
 
ProConnex® Standard Flow Path for MidiKros or MidiKros TC
  SPECSTUBEGN15390S
ProConnex® Standard Flow Path for MidiKros or MidiKros TC
  SPECSTUBEGN15390S

 

  • Description:
    Hollow fiber, MidiKros or midiKros TC, 250 ml
  • Type:
    Conical
  • Pk:
    1

 

 

ProConnex® standard flow path for midiKros or midiKros TC, 250 ml, 3/Pk.

System: KR2i; pressor sensors: luer; delivery condition: sterile; compatible with: MidiKros (D) or MidiKros TC (T).

HUV recirculation: 3,1 ml; HUV reservoir: 1 ml; HUV total tubeset: 34,7 ml.

 : Filter sold separately.