ContactUsLinkComponent
Protein quantitation assay kits, Non-Interfering™ (NI™) Protein Assay
  786-005
 :  
undefined
Protein quantitation assay kits, Non-Interfering™ (NI™) Protein Assay
  786-005
 :  
  • Pk:
    1 KIT
  • Description:
    Non-Interfering™ (NI™) Protein Assay with albumin standard, 500 assays
A highly sensitive colorimetric protein assay that overcomes interference of common laboratory agents present in protein solutions and shows minimal protein-to-protein variation. The assay is unaffected by the presence of common laboratory agents such as reducing agents, chelating agents, detergents, amines, sugars, chaotropes, salts, drugs, antibiotics, cobalt, and others.