Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pipette Tip Boxes and Racks
Support Frame for TipBox
Support Frame for TipBox
Catalog # 613-7260
Supplier:
Brand
Expand image
Support Frame for TipBox
Catalog # 613-7260
Supplier:
Brand
Specifications
Description:
Support frame for TipBox 1250 µl, material: PC/PBT
Pk:
1
Specifications
About this item
Product Details
Product Details