ContactUsLinkComponent
Configurator for Gas Detection System, X-am® 2800
Configurator for Gas Detection System, X-am® 2800
  DRAE3703900
 :  Dräger
Configurator for Gas Detection System, X-am® 2800
  DRAE3703900
 :  Dräger
  • Description:
    Drager X-am 2800 configurator
  • Pk:
    1
Configurator for Dräger X-am® 2800 consisting of basic device with data logger, delivered with manufacturer and calibration certificate.
  • Configurator: up to 4 sensors can be selected, sensor configuration can be changed later in product life
Customer-specific settings (firmware) can be selected when ordering, examples: Measurement gas for the Ex sensor (factory setting: methane), alarm thresholds, alarm behaviour, quick menu options, D-light etc.
 : Power supply (battery) optional (factory setting: with battery), without charger.