Order Entry
Austria
ContactUsLinkComponent
DrySyn® MAXI Heating Blocks, 2 L
DrySyn® MAXI Heating Blocks, 2 L
  ASYNADS2-2
 :  ASYNT
DrySyn® MAXI Heating Blocks, 2 L
  ASYNADS2-2
 :  ASYNT

 

  • Description:
    Heating blocks, 2 L with heat resistant safety lifting handles
  • Environmentally Preferable:
  • Pk:
    1

 

 

The DrySyn® MAXI 2 L is a stand-alone 2000 ml version of the standard DrySyn MAXI Kit (which alternatively has a 3000 ml base and 2000 ml insert) which comes with heat resistant safety lifting handles.

  • Enables clean, safe synthesis without the hazards or problems associated with silicon oil baths or heating mantles
  • With no messy oil, heat resistant handles, and simple to configure range of inserts this system is world renowned
  • With heating rates the same as an oil bath there is no need to change your chemistry either
  • Fits standard hotplate stirrers with ease

Does not require any further base, and is suitable for heating 2000 ml round bottom flasks.