Order Entry
Austria
ContactUsLinkComponent
Cuvettes for Spectrophotometers
Cuvettes for Spectrophotometers
Catalog # 634-9121
Supplier:  HELLMA
undefined
Cuvettes for Spectrophotometers
Catalog # 634-9121
Supplier:  HELLMA

Specifications

  • Path length:
    5 mm
  • Type:
    Semi-micro
  • Volume:
    700 µl
  • Window material:
    Quartz (Suprasil®)
  • Height:
    45 mm
  • Pk:
    1
  • Base thickness:
    3,2 mm

Specifications

About this item

Quartz (Suprasil®) cuvette for spectroscopy applications.

Suitable for high precision analytical techniques.