ContactUsLinkComponent
Kits: whose DNA was left behind?
Kits: whose DNA was left behind?
Catalog # 763-0008
Supplier:  EDVOTEK
undefined
Kits: whose DNA was left behind?
Catalog # 763-0008
Supplier:  EDVOTEK
Specifications
  • Description:
    Whose DNA Was Left Behind?
  • Pk:
    1 KIT
Specifications
About this item
DNA obtained from a single hair left behind at a crime scene can be used to identify a criminal. In this experiment, students will compare simulated crime scene DNA with that of two suspects.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Delivery: Kit includes instructions, Ready-to-Load™ dye sample, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.