ContactUsLinkComponent
Experimentation kits, DNA fingerprinting - using PCR
Experimentation kits, DNA fingerprinting - using PCR
  EDVO371
 :  EDVOTEK
undefined
Experimentation kits, DNA fingerprinting - using PCR
  EDVO371
 :  EDVOTEK
  • Description:
    DNA Fingerprinting - Using PCR
  • Pk:
    1 KIT
Students can solve a crime using PCR. Plasmid DNA is provided that, when amplified by PCR, provides products that represent individual DNA profiles.
  • For 6 lab groups
  • Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
 : Kit includes instructions, PCR beads, DNA templates, primers, DNA ladder, ultrapure water, wax beads, agarose, loading dye, electrophoresis buffer, and gel stain.