ContactUsLinkComponent
Experimentation kits, principles of enzyme catalysis
Experimentation kits, principles of enzyme catalysis
Catalog # EDVO282
Supplier:  EDVOTEK
undefined
Experimentation kits, principles of enzyme catalysis
Catalog # EDVO282
Supplier:  EDVOTEK
Specifications
  • Description:
    Principles of Enzyme Catalysis
  • Pk:
    1 KIT
Specifications
About this item
This easy and safe experiment allows students to learn about enzyme catalysis, the nature of enzyme action, and protein structure-function relationship. Students will perform an enzyme assay and determine the rate of the enzymatic reaction.
  • For 10 lab groups
  • Requires 30 to 45 minutes
Also required: Visible wavelength spectrophotometer; 1 ml, 5 ml, and 10 ml pipettes; and linear graph paper.
Delivery: Kit includes instructions, catalase solution, hydrogen peroxide, phosphate buffer, assay reagent, acidification solution, colour enhancer and developer.