ContactUsLinkComponent
Experimentation kits, simulation of HIV detection by western blot (agarose-based)
Experimentation kits, simulation of HIV detection by western blot (agarose-based)
Catalog # EDVO275
Supplier:  EDVOTEK
undefined
Experimentation kits, simulation of HIV detection by western blot (agarose-based)
Catalog # EDVO275
Supplier:  EDVOTEK
Specifications
  • Description:
    Simulation of HIV Detection by Western Blot (Agarose-based)
  • Pk:
    1
Specifications
About this item
The second assay used to confirm a positive HIV ELISA result is the Western blot. Using this kit, students separate protein samples from hypothetical patients on agarose gels. The proteins are then transferred to a membrane and simulated HIV proteins are detected.
  • For 6 lab groups
  • Electrophoresis 45 minutes; blot overnight; detection 25 minutes
Also required: Electrophoresis tank, power supply, isopropanol, glacial acetic acid, and 37 °C incubator.
Delivery: Kit includes instructions, positive and negative controls, simulated patient samples, standard molecular weight markers, protein agarose, buffer, PVDF membrane, protein stain, filter paper, practice gel loading solution.