ContactUsLinkComponent
Experimentation kits, principles of thin layer chromatography
Experimentation kits, principles of thin layer chromatography
Catalog # EDVO113
Supplier:  EDVOTEK
undefined
Experimentation kits, principles of thin layer chromatography
Catalog # EDVO113
Supplier:  EDVOTEK
Specifications
  • Description:
    Principles of Thin Layer Chromatography
  • Pk:
    1
Specifications
About this item
This experiment introduces chromatographic theory and methods of thin layer chromatography (TLC). A mixture of dyes are separated on a cellulose-based TLC plate using different solvent systems.
  • For 8 lab groups
  • Spotting plates 20 minutes, TLC separation 5 minutes
Also required: 5ml or 10 ml pipettes.
Delivery: Kit includes instructions, samples, reagents and solvents, cellulose TLC plate, 5 µl capillary pipettes.