ContactUsLinkComponent
Experimentation kits, how clean is the water we drink and the air we breathe?
Experimentation kits, how clean is the water we drink and the air we breathe?
Catalog # EDVOS-30
Supplier:  EDVOTEK
undefined
Experimentation kits, how clean is the water we drink and the air we breathe?
Catalog # EDVOS-30
Supplier:  EDVOTEK
Specifications
  • Description:
    ReadyPour™ Luria Broth Agar Base
  • Pk:
    1
Specifications
About this item
With this kit, students sample water and air and then grow any microbes present overnight. A safe and simple way to teach pollution.
  • For 10 lab groups
  • Complete in 30 minutes and grow overnight
Also required: Microscope with 400× magnification.
Delivery: Kit includes instructions, multispot slides (4 cell types each), eosin and methylene blue stain, mounting medium, cover slips, transfer pipettes, and immersion troughs.