ContactUsLinkComponent
Experimentation kits, what is an epidemic and how does an infection spread?
Experimentation kits, what is an epidemic and how does an infection spread?
Catalog # EDVOS-68
Supplier:  EDVOTEK
undefined
Experimentation kits, what is an epidemic and how does an infection spread?
Catalog # EDVOS-68
Supplier:  EDVOTEK
Specifications
  • Description:
    What is an Epidemic and How Does An Infection Spread?
  • Pk:
    1
Specifications
About this item
Infectious agents such as bacteria and viruses can spread rapidly through a population and cause widespready disease and death. In this experiment, students use coloured solutions to simulate the spreading of a disease in the classroom.
  • For 10 lab groups
  • Complete in 30 minutes
Delivery: Kit includes instructions, HCl solution, NaOH, colour indicator, test tubes, and pipettes.