ContactUsLinkComponent
Experimentation kits, introduction to ELISA reactions
Experimentation kits, introduction to ELISA reactions
  EDVO269
 :  EDVOTEK
Experimentation kits, introduction to ELISA reactions
  EDVO269
 :  EDVOTEK
  • Description:
    Introduction to ELISA Reactions
  • Pk:
    1
Students learn the basic principles of the Enzyme-linked Immunosorbent Assay (ELISA) using this precise and sensitive antibody-based detection kit.
  • For 10 Lab Groups
  • Complete in 45 minutes
Also required: Distilled or deionised water, 37 °C incubator, automatic micropipettes with tips, and laboratory glassware.
 : Kit includes instructions, antigens, primary and secondary antibodies, peroxide co-substrate, hydrogen peroxide, ABTS substrate, phosphate buffered saline, tubes, plates, and transfer pipettes.