ContactUsLinkComponent
Kits: the case of the invisible bands, Ready-to-Load™
Kits: the case of the invisible bands, Ready-to-Load™
Catalog # 763-0009
Supplier:  EDVOTEK
undefined
Kits: the case of the invisible bands, Ready-to-Load™
Catalog # 763-0009
Supplier:  EDVOTEK
Specifications
  • Description:
    The Case of the Invisible Bands
  • Pk:
    1
Specifications
About this item
Experiment using fluorescence and electrophoresis to solve the mystery of the invisible bands.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank, power supply and long wave UV lamp.
Delivery: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.