ContactUsLinkComponent
Ready-to-Load™ Kits, what is PCR and how does it work?
  EDVOS-48
 :  EDVOTEK
undefined
Ready-to-Load™ Kits, what is PCR and how does it work?
  EDVOS-48
 :  EDVOTEK
  • Description:
    What is PCR and How Does it Work?
  • Pk:
    1 KIT
This simulation experiment demonstrates the process of DNA amplification by PCR and how the amplified product is detected by separating the reaction mixture by agarose gel electrophoresis.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
 : Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.