ContactUsLinkComponent
What does DNA look like?
What does DNA look like?
Artikelnr. EDVOS-10
Lieferant:  EDVOTEK
undefined
What does DNA look like?
Artikelnr. EDVOS-10
Lieferant:  EDVOTEK
Spezifikationen
  • Description:
    What Does DNA Look Like?
  • Pk:
    1
Spezifikationen
Über diesen Artikel
This Lab activity shows students what real chromosomal DNA Looks Like and allows them to explore the procedures involved in DNA extraction.
  • For 10 lab groups
  • Complete in 30 minutes
Also required: 95% ethanol or isopropyl alcohol.
Delivery: Kit includes instructions, DNA extraction buffer, DNA sample in capped test tube, transfer pipettes, minilinks, glass rod, DNA spooling rods, test tubes, and salt.