ContactUsLinkComponent
Experimentation kits, PCR-based VNTR human DNA typing
Experimentation kits, PCR-based VNTR human DNA typing
Catalog # EDVO334
Supplier:  EDVOTEK
undefined
Experimentation kits, PCR-based VNTR human DNA typing
Catalog # EDVO334
Supplier:  EDVOTEK
Specifications
  • Description:
    PCR-based VNTR Human DNA Typing
  • Pk:
    1 KIT
Specifications
About this item
In DNA fingerprinting, variable number tandem repeats (VNTR) are used to identify individuals. Using this kit, students type themselves at the D1S80 locus on chromosome 1. This region contains between 14 and 40 copies of a 16 base pair repeat.
  • For 25 students
  • Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
Delivery: Kit includes instructions, proteinase K, PCR beads, control DNA and primers, microtubes, chelating agent, agarose, DNA ladder, practice gel loading solution, gel loading dye, electrophoresis buffer, and gel stain.