Orders LinkContactUsLinkComponent
Experimentation kits, mitochondrial DNA analysis using PCR
Experimentation kits, mitochondrial DNA analysis using PCR
Mitochondrial DNA Analysis Using PCR
Catalog # EDVO332
Supplier:   EDVOTEK
Experimentation kits, mitochondrial DNA analysis using PCR
Catalog # EDVO332
Supplier:  EDVOTEK
Experimentation kits, mitochondrial DNA analysis using PCR
Experimentation kits, mitochondrial DNA analysis using PCR
Mitochondrial DNA Analysis Using PCR
Price
132,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
The mitochondria are thought to have evolved from a symbiotic relationship between prokaryotic and eukaryotic cells. Mitochondria have their own DNA and are only inherited via the maternal line. In this experiment, students amplify two regions of their mitochondrial DNA.
  • For 25 students
  • Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
Delivery information: Kit includes instructions, proteinase K, PCR beads, control DNA and primers, microtubes, chelating agent, agarose, DNA ladder, practice gel loading solution, gel loading dye, electrophoresis buffer, and gel stain.
Specifications
  • Description:
    Mitochondrial DNA Analysis Using PCR