ContactUsLinkComponent
Experimentation kits, amplification of DNA by PCR
Experimentation kits, amplification of DNA by PCR
Catalog # 763-0001
Supplier:  EDVOTEK
undefined
Experimentation kits, amplification of DNA by PCR
Catalog # 763-0001
Supplier:  EDVOTEK
Specifications
  • Description:
    Amplification of DNA by PCR
  • Pk:
    1 KIT
Specifications
About this item
In this PCR experiment, students make billions of copies of a small amount of DNA in only 90 minutes. They just need to mix template DNA and primers with PCR beads that contain all of the other components required to carry out a PCR reaction. Students see the increasing amounts of DNA for themselves, taking samples every few cycles and analysing them on a DNA gel.
  • For 10 lab groups
  • PCR 3 hours; electrophoresis 45 minutes
Also required: 5 to 50 µl adjustable micropipettes, tips, thermal cycler, electrophoresis gel tank and power supply.
Delivery: Kit includes instructions, PCR beads, DNA template and primers, DNA size ladder, ultrapure water, wax beads, gel loading dye, agarose, electrophoresis buffer, and gel stain.