ContactUsLinkComponent
Experimentation kits, isolation of E. coli Chromosomal DNA
Experimentation kits, isolation of E. coli Chromosomal DNA
Catalog # EDVO203
Supplier:  EDVOTEK
undefined
Experimentation kits, isolation of E. coli Chromosomal DNA
Catalog # EDVO203
Supplier:  EDVOTEK
Specifications
  • Description:
    Isolation of E. coli Chromosomal DNA
  • Pk:
    1 KIT
Specifications
About this item
Isolation of high molecular weight chromosomal DNA is the first step in molecular cloning since it is the source of genes in cells. This experiment provides DNA extraction LyphoCells and reagents for isolating chromosomal DNA from E. coli. After spooling from solution, the DNA can be dissolved and analysed by agarose gel electrophoresis as an optional lab extension activity.
  • For 5 lab groups
  • Complete in 60 minutes
Also required: Water bath, 95 to 100% isopropanol. For optional electrophoresis: Electrophoresis tank, power supply, 5 to 50 µl adjustable or 40 µl fixed volume micropipettes, and white light box.
Delivery: Kit includes instructions, LyphoCells, various solutions and buffers, agarose powder, and stains.