Order Entry
Austria
Orders LinkContactUsLinkComponent
Carousel tissue processors

Carousel tissue processors

Robust carousel tissue processor, MTP, 10...35 °C, 110/240 V, 50/60 Hz, 850×850×750 mm, 80 kg
Catalog # SLEE11000200
Supplier:   Slee
Carousel tissue processors
Catalog # SLEE11000200
Supplier:  Slee
Carousel tissue processors
Carousel tissue processors

Robust carousel tissue processor, MTP, 10...35 °C, 110/240 V, 50/60 Hz, 850×850×750 mm, 80 kg

Price
22 400,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The robust carousel tissue processor guarantees gentle and highly reliable specimen processing in conjunction with state of the art control features. The specimen throughput can be doubled to 240 cassettes per run with a second transport basket. For faster and substantially improved processing of tissue, the instrument can optionally be equipped with vacuum units for all paraffin beakers. An integrated battery ensures highest process safety even in environments with frequent power failures. Hazardous fumes are efficiently eliminated by a fume extraction system thereby increasing user safety. The system is easy loading and has a full process control by rotating table principle.

  • Easy loading and full process control by rotating table principle
  • Integrated UPS system protects your samples
  • Intuitive control panel
  • Delayed start function
  • System status monitor
  • Password protected software
  • Power draw of about 250 VA, and I protective class along with 2× T 6,3 A power fuses

Maximum heat emission of about 250 J/s, temperature range during storage is 5 to 55 °C, relative humidity is maximum 80% non condensing along with pressure difference 0,5 bar maximum (500 hPa).

Delivery information: System includes operation manual, 10× glass beakers (DURAN®), 2× heated stainless steel paraffin beakers, 1× stainless steel transport basket, capacity of 120 standard cassettes, mains cable.

Specifications
  • Temperature Range:
    10...35 °C
  • Weight:
    80 kg
  • Description:
    Robust carousel tissue processor, MTP
  • W×D×H:
    850×850×750 mm
  • Electrical supply:
    110/240 V, 50/60 Hz
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.