ContactUsLinkComponent
MagSorb-16 Magnetic Rack for Manual Nucleic Acid Extraction
MagSorb-16 Magnetic Rack for Manual Nucleic Acid Extraction
Catalog # BSANBS-010601
Supplier:  BIOSAN
MagSorb-16 Magnetic Rack for Manual Nucleic Acid Extraction
Catalog # BSANBS-010601
Supplier:  BIOSAN
Specifications
  • No. of Holes:
    16
  • Description:
    MagSorb-16 Magnetic rack
  • Pk:
    1
Specifications
About this item
Biosan presents the complete line of necessary instrumentation to utilise magnetic bead extraction kits and protocols from various manufacturersand meet the most demanding user requirements.
The one of the foundations of this line is MagSorb-16 which is a magnetic rack that easily accommodates up to 16 single usetubes (1,5 to 2 ml). Therack consists of following parts: tube mounting racksand magnetic stand. Magnetic beads based NA extraction methods offer quick and efficient separation of genetic material from cellular leftovers.