ContactUsLinkComponent
HLP Medium (Hsu's Lactobacillus/Pediococcus Medium)
HLP Medium (Hsu's Lactobacillus/Pediococcus Medium)
Catalog # DOEH923556244
Supplier:  DOEHLER GROUP
CAS Number:  
undefined
HLP Medium (Hsu's Lactobacillus/Pediococcus Medium)
Catalog # DOEH923556244
Supplier:  DOEHLER GROUP
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    1
  • Description:
    HLP Medium (Hsu's Lactobacillus/Pediococcus Medium)
  • Filling:
    500 g
Specifications
About this item
Siebel-Instituts nutrient medium for the selective detection and quantitative determination of Lactobacillus and Pediococcus from the brewing environment.
  • Certified by the Siebel-Institute for technology
  • Ready-to-use powder to prepare HLP gel or agar without prior autoclaving
  • Functionally tested with strains originally isolated from the brewing environment
HLP promote the growth of beer spoiling bacteria like Lactobacillus spp./Pedicococcus spp. which are commonly encountered in a brewery. Cycloheximide contained in the medium inhibits the growth of culture yeast.