Express Eingabe
Austria
AuftragsverfolgungKontakt
Containers with Sloping Base
Containers with Sloping Base
Containers with Sloping Base
Containers with Sloping Base
Containers with Sloping Base
These drums which incorporate a sloping base allow the container to drain completely.
  • Made from 316L stainless steel
  • Includes lid, clamp and silicone seal
  • Other seals available
  • Ferrule for easy emptying
  • Handles enable easy transportation
The Sampling Systems range of containers with sloping base ensures that material can be completely drained from within and include a ferrule at the base for easy emptying. They have a mirror polish finish for easy cleaning.
Product Family Options
  • Bezeichnung
    Kapazität
    Material
  • Container with sloping base
    100 L
    Edelstahl 316L
    1 * 1 ST
    1 * 1 ST - 2 520,00 €
    2 520,00 €
    1 * 1 ST
       2 520,00 €
  • Container with sloping base
    200 L
    Edelstahl 316L
    1 * 1 ST
    1 * 1 ST - 3 290,00 €
    3 290,00 €
    1 * 1 ST
       3 290,00 €
  • Container with sloping base
    50 L
    Edelstahl 316L
    1 * 1 ST
    1 * 1 ST - 2 260,00 €
    2 260,00 €
    1 * 1 ST
       2 260,00 €
  • 90° bend
    Edelstahl 316L
    1 * 1 ST
    1 * 1 ST - 222,00 €
    222,00 €
    1 * 1 ST
       222,00 €
  • Hygienic valve kit with EPDM seals
    1 * 1 ST
    1 * 1 ST - 464,00 €
    464,00 €
    1 * 1 ST
       464,00 €
  • Hygienic valve kit with silicone seals
    1 * 1 ST
    1 * 1 ST - 476,00 €
    476,00 €
    1 * 1 ST
       476,00 €