Orders LinkContactUsLinkComponent
Pointed Scissors
Pointed Scissors
Pointed Scissors
Pointed Scissors
Surgical grade scissors for cutting.
  • Made from surgical grade 420 stainless steel
  • Great for cutting and manipulating samples
These high quality pointed scissors are made from surgical grade 420 stainless steel and great for cutting and manipulating.
Product Family Options
  • Length
  • 150 mm
    1 * 1 ST
    1 * 1 ST - 10,60 €
    10,60 €
    1 * 1 ST
       10,60 €
  • 310 mm
    1 * 1 ST
    1 * 1 ST - 37,90 €
    37,90 €
    1 * 1 ST
       37,90 €