ContactUsLinkComponent
Stainless Test Tube Racks
Stainless Test Tube Racks
Catalog # SAMPA645-12
Supplier:  Sampling Systems
undefined
Stainless Test Tube Racks
Catalog # SAMPA645-12
Supplier:  Sampling Systems
Specifications
  • Description:
    Test tube rack for 12 test tubes
  • Material:
    Stainless steel 304
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Test tube racks made from 304 stainless steel to hold test tubes.
  • Made from corrosion resistant 304 stainless steel
  • Ideal for use in laboratories and production
  • Mirror polish finish
  • Various sizes available
These stainless steel test tube racks come in two standard sizes to securely hold 12 and 48 test tubes and are made from 304 stainless steel which mean they are ideal for GMP applications. Other sizes are available on request.